Cryoboxes, 136×136 mm
Leverantör: TENAK
Certifikat
Om denna produkt
Cryo resistant fibreboard/cardboard.
- Water repellent for longer durability
- Various combinations possible with different dividers (to be ordered separately)
- Available in different heights and colours
For storage of cryovials at low temperatures.
*Height stated is the interior height.