Sheep Cyclooxygenase (from Seminal vesicles)
Katalog # ENZOBMLEH0010001
Leverantör: ENZO LIFE SCIENCES
CAS-nummer:
Specifikationer
- Pk:1 mg
- Source:Seminal vesicles
- Species:Sheep
- Seq:02
- Form:Liquid
- Enzyme name:Cyclooxygenase
- Purity:>95%
- Formulation:In 100 mM TRIS/HCl, pH 8,0, containing 1% Tween 80.
Specifikationer
Om denna produkt
Isolated from ram seminal vesicles.