Order Entry
Sweden
ContactUsLinkComponent
Mouse recombinant TGF-beta3
Mouse recombinant TGF-beta3
Supplier:  ENZO LIFE SCIENCES
Certificates
Mouse recombinant TGF-beta3
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Mouse
  • Biological activity:
    The activity is determined by the cell toxicity assay, usi ng the WHO Standard 98/608 as a direct comparison, which is typically observed at concentations <0,05 ng/ml.
  • Protein synonyms:
    Transforming growth factor beta3
  • Protein/peptide name:
    TGF-beta3
  • Purity:
    >98% (Reducing and Non-reducing SDS-PAGE)
  • Molecular weight:
    ~25,5kDa
  • Concentration:
    0,25 mg/ml
  • Formulation:
    Liquid. In 20% ethanol and 0,12% acetic acid.

Specifications