Order Entry
Sweden
ContactUsLinkComponent
Human recombinant SUMO activating enzyme E1 (from E. coli), His-tag
Human recombinant SUMO activating enzyme E1 (from E. coli), His-tag
Catalog # ENZOBMLUW93300025
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Human recombinant SUMO activating enzyme E1 (from E. coli), His-tag
Catalog # ENZOBMLUW93300025
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    25 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Seq:
    02
  • Tag sequence:
    HIS Tag
  • Protein/peptide name:
    SUMO activating enzyme E1
  • Concentration:
    0,5 mg/ml
  • Formulation:
    Liquid. In HEPES, pH 8,0, containing 110 mM potassium acetate, 2 mM magnesium acetate, 0,5 mM EGTA and 1 mM DTT.

Specifications

About this item

Produced in E. coli. Full length heterodimeric human SUMO-activating enzyme.