Order Entry
Sweden
ContactUsLinkComponent
Bovine native Calmodulin (from Brain), Agarose
Bovine native Calmodulin (from Brain), Agarose
Catalog # ENZOBMLSE1020001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Bovine native Calmodulin (from Brain), Agarose
Catalog # ENZOBMLSE1020001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Conjugation:
    Agarose
  • Protein/peptide type:
    Native
  • Source:
    Brain
  • Species:
    Bovine
  • Seq:
    02
  • Protein/peptide name:
    Calmodulin
  • Concentration:
    Calmodulin concentration is 1 mg/ml beads
  • Formulation:
    Liquid. Suspension.

Specifications

About this item

Isolated from bovine brain. Calmodulin conjugated to 4% cross-linked agarose beads via a 5 atom hydrophilic spacer arm.