SAMS peptide (phosphorylated)
Catalog # ENZOBMLP2330001
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 mg
- Conjugation:Biotin
- Seq:02
- Protein/peptide name:SAMS Peptide (phosphorylated)
- Purity:95%
Specifications
About this item
AMPK substrate
This peptide, biotin-HMRSAMpSGLHLVKRR, contains a biotin group at its amino terminus and a phospho-serine. It is derived from rat acetyl-CoA carboxylase. It may be used as a control peptide for unphosphorylated SAMS Peptide (Prod. No. BML-P231 and BML-P232). The biotin allows peptide to be used in kinase assays with streptavidin-bound membranes.