Ac-IEPD-pNA
Catalog # ENZOBMLP1330005
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:5 mg
- Seq:02
- Protein/peptide name:Ac-IEPD-pNA
- Purity:95%
Specifications
About this item
Caspase substrate
Colorimetric granzyme B substrate. Combinatorial and kinetic studies have found the IEPD sequence to be optimal for cleavage of a tetrapeptide substrate by recombinant granzyme B, with a Km for Ac-IEPD-pNA of 57 µM. Cleavage of the substrate by granzyme B releases pNA, which can be detected by its absorbance at 405 nm.