Recombinant DsbC (from E. coli)
ENZOALX201268C100
New Product
:
- Pk:100 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:
- Seq:02
- Environmentally Preferable:
- Protein/peptide name:DsbC
- Purity:>95% (SDS-PAGE)
- Concentration:1 mg/ml
- Formulation:Liquid. In 20 mM TRIS, pH 7,5, containing 2 mM EDTA.
Produced in E. coli.