Order Entry
Sweden
ContactUsLinkComponent
ORGANELLE-ID-RGB® III Assay kit
ORGANELLE-ID-RGB® III Assay kit
Catalog # ENZOENZ51032K100
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
ORGANELLE-ID-RGB® III Assay kit
Catalog # ENZOENZ51032K100
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Description:
    ORGANELLE-ID-RGB® III assay kit (100 assays)
  • Seq:
    02
  • Storage conditions:
    –20 °C (Short term), –80 °C (Long term)
  • Application:
    Fluorescence microscopy, fluorescent detection

Specifications

About this item

TheORGANELLE-ID-RGB® III assay kit contains Golgi apparatus-selective, endoplasmic reticulum-selective and nucleus-selective dyes suitable for live cell staining.

  • Highly resistant to photo-bleaching, concentration quenching and photoconversion
  • Identify modulators of secretory membrane traffic and organelle integrity

The green dye component of the Organelle-ID RGB™ III detection reagent is more faithfully localized to the Golgi apparatus, with minimal staining of the endoplasmic reticulum. The red dye component of the reagent stains the endoplasmic reticulum with high fidelity and is specifically designed for use with green fluorescing probes.

ORGANELLE-ID-RGB® III assay kit is validated with human cervical carcinoma cell line HeLa, human T-lymphocyte cell line, Jurkat, canine kidney cell line MDCK, and human bone osteosarcoma epithelial cell line U2OS. The kit should also be suitable for identifying Golgi body and endoplasmic reticulum perturbing agents and thus can be a useful tool for examining the transport and recycling of molecules from the GA to ER in cellular secretory pathways.

Delivery: Kit includes ORGANELLE-ID-RGB® Reagent III (lyophilized) (1 vial), 10X Assay Buffer 1 (15 ml), 50X Assay Buffer 2 (1.2 ml).