Anti-TRAIL-R2 Goat Polyclonal Antibody
ENZOALX210743C200
- Antibody type:Primary
- Antigen name:TNF Receptor superfamily member 10b
- Antigen symbol:TRAIL-R2
- Clonality:Polyclonal
- Flow cytometry:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Western blot:Yes
- Seq:02
- Form:Liquid. In PBS containing 1 mg/ml BSA and 0,1% sodium azide.
- Antigen synonyms:TRAILR2|TRAIL-R2|CD262|Trail receptor 2|TRICK2|KILLER|TRICK2A|TRICKB|ZTNFR9|KILLER/DR5|TRICK2B|DR5
- Concentration:1 mg/ml
- Purification:Epitope-affinity purified IgG
- Pk:200 µG
Host: Goat
- For Immunocytochemistry, Immunohistochemistry, Western Blot
Recognizes human and mouse TRAIL-R2. Detects a band of ~52kDa by Western blot.
Type: Primary
Antigen: TRAIL-R2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human