Anti-Ubiquitin Rabbit Polyclonal Antibody
Leverantör: ENZO LIFE SCIENCES
Certifikat
Om denna produkt
Ubiquitin (Ub) plays a very important role in regulated non-lysosomal ATP dependent protein degradation. The protein to be degraded is conjugated to Ub and the ubiquinated protein is then selectively degraded by the 26S complex, a multicatalytic cytosolic and nuclear protease. The Ub-proteasome proteolytic pathway, which is a complex process, is implicated to be of great importance for regulating numerous cellular processes.
- For Immunocytochemistry, Western Blot
Recognizes human, mouse, rat, bovine, dog, chicken, Drosophila, fish, monkey, hamster, rabbit, guinea pig, pig, sheep, Xenopus and yeast Ubiquitin. Detects a band of ~10kDa by Western blot.
Type: Primary
Antigen: UBIQUITIN
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Specifikationer
- Catalog No:
- ENZOADISPA200F
- ENZOADISPA200D
- Antigen symbol:
- Ubi-1
- Ubi-1
- Antigen name:
- Ubiquitin
- Ubiquitin
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Host:
- Rabbit
- Rabbit
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid. In PBS, pH 7,2, containing 50% glycerol and 0,09% sodium azide.
- Liquid. In PBS, pH 7,2, containing 50% glycerol and 0,09% sodium azide.
- Purification:
- Protein A affinity purified
- Protein A affinity purified