Order Entry
Sweden
ContactUsLinkComponent
 
(2-Nitrophenyl)-β-D-galactopyranoside (ONPG) ≥99% for molecular biology
Catalog # USBIO7000
Supplier:  US Biological
CAS Number:  
(2-Nitrophenyl)-β-D-galactopyranoside (ONPG) ≥99% for molecular biology
Catalog # USBIO7000
Supplier:  US Biological
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to slight yellow powder
  • Purity:
    ≥99%
  • Identity (IR):
    Conforms to reference
  • Solubility (1%, H₂O):
    Colorless to slight yellow, clear, complete DMSO (slightly), water (slightly, sonicated)
  • Turbidity (1%, H₂O):
    ≤4 NTU
  • Water (KF):
    ≤0,5%
  • Specific rotation (C=1, H₂O):
    [a]D: −68,0 ± 3°
  • Free o-nitrophenol:
    ≤0,03%
  • Bromine:
    ≤2300 mg/kg
  • Aluminum:
    ≤560 mg/kg

Specification Test Results

About this item

 :
 :301.25 g/mol
Storage Temperature:Refrigerator
CAS Number:369-07-3
Formula:C₁₂H₁₅NO₈
MDL Number:MFCD00063255