Order Entry
Sweden
ContactUsLinkComponent
Sodium poly(naphthaleneformaldehyde)sulfonate
Sodium poly(naphthaleneformaldehyde)sulfonate
Catalog # SPCMS1161-100GM
Supplier:  Spectrum Chemical
CAS Number:  
Sodium poly(naphthaleneformaldehyde)sulfonate
Catalog # SPCMS1161-100GM
Supplier:  Spectrum Chemical
CAS Number:  

Specifications

  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Appearance:
    Beige powder

Specification Test Results

About this item

 :
CAS Number:9084-06-4
Formula:C₁₀H₈O₃S·CH₂O)n·xNa