Order Entry
Sweden
ContactUsLinkComponent
Organic reference standard, fluthiacet-methyl, Dr. Ehrenstorfer
Organic reference standard, fluthiacet-methyl, Dr. Ehrenstorfer
Catalog # EHERC13862000
Supplier:  EHRENSTORFER
CAS Number:  
Organic reference standard, fluthiacet-methyl, Dr. Ehrenstorfer
Catalog # EHERC13862000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pk:
    0,1 g
  • Standards:
    Reference standard
  • CAS No.:
    117337-19-6
  • Description:
    Fluthiacet-methyl
  • Formula:
    C₁₅H₁₅ClFN₃O₃S₂
  • Description:
    Fluthiacet-methyl

Specifications

About this item