Order Entry
Sweden
ContactUsLinkComponent
 
Anti-VEGF Polyclonal Antibody
Catalog # BSBTPB9071
Supplier:  Boster Bio
undefined
Anti-VEGF Polyclonal Antibody
Catalog # BSBTPB9071
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    VEGF
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Rat
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Polyclonal antibody for VEGF/VEGFA detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. VEGF/VEGFA information: Molecular Weight: 27042 MW; Subcellular Localization: Secreted . VEGF121 is acidic and freely secreted. VEGF165 is more basic, has heparin-binding properties and, although a signicant proportion remains cell-associated, most is freely secreted. VEGF189 is very basic, it is cell-associated after secretion and is bound avidly by heparin and the extracellular matrix, although it may be released as a soluble form by heparin, heparinase or plasmin; Tissue Specificity: Isoform VEGF189, isoform VEGF165 and isoform VEGF121 are widely expressed. Isoform VEGF206 and isoform VEGF145 are not widely expressed. A higher level expression seen in pituitary tumors as compared to the pituitary gland.

Type: Primary
Antigen: VEGF
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Rat