Anti-SCP3 Polyclonal Antibody
Catalog # BSBTRP1035
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen symbol:SCP3
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Polyclonal antibody for SCP3/SYCP3 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Human. SCP3/SYCP3 information: Molecular Weight: 27729 MW; Subcellular Localization: Nucleus . Chromosome . In tripartite segments of synaptonemal complexes, irrespective of whether these are synapsed or unsynapsed; Tissue Specificity: Testis-specific.
Type: Primary
Antigen: SCP3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human