Anti-PD-1 Polyclonal Antibody
Catalog # BSBTRP1039
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen symbol:PD-1
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human,Rat
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Polyclonal antibody for PD 1/PDCD1 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Human. PD 1/PDCD1 information: Molecular Weight: 31647 MW; Subcellular Localization: Membrane; Single-pass type I membrane protein.
Type: Primary
Antigen: PD-1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Rat