Anti-KLF6 Polyclonal Antibody
BSBTPB9231
- Antibody type:Primary
- Antigen symbol:KLF6
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human,Rat
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Polyclonal antibody for KLF6 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Human. KLF6 information: Molecular Weight: 31865 MW; Subcellular Localization: Nucleus; Tissue Specificity: Highly expressed in placenta followed by spleen, thymus, prostate, testis, small intestine and colon. Weakly expressed in pancreas, lung, liver, heart and skeletal muscle. Also expressed in fetal brain, spleen and thymus.
Type: Primary
Antigen: KLF6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Rat