Anti-KCNMA1 Polyclonal Antibody
Catalog # BSBTPB9227
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen symbol:KCNMA1
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Polyclonal antibody for KCNMA1 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. KCNMA1 information: Molecular Weight: 137560 MW; Subcellular Localization: Cell membrane ; Multi-pass membrane protein ; Tissue Specificity: Widely expressed. Except in myocytes, it is almost ubiquitously expressed.
Type: Primary
Antigen: KCNMA1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat