Order Entry
Sweden
ContactUsLinkComponent
 
Anti-IL-15 Polyclonal Antibody
  BSBTPB9244
undefined
Anti-IL-15 Polyclonal Antibody
  BSBTPB9244

 

  • Antibody type:
    Primary
  • Antigen symbol:
    IL-15
  • Clonality:
    Polyclonal
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Polyclonal antibody for IL-15/IL15 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: ELISA. Reactive species: Human. IL-15/IL15 information: Molecular Weight: 18086 MW; Subcellular Localization: Isoform IL15-S48AA: Secreted; Tissue Specificity: Most abundant in placenta and skeletal muscle. It is also detected in the heart, lung, liver and kidney. IL15- S21AA is preferentially expressed in tissues such as testis and thymus.

Type: Primary
Antigen: IL-15
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human