Anti-IKB Beta Polyclonal Antibody
Catalog # BSBTPB9292
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen symbol:IKB Beta
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Polyclonal antibody for IKB BETA/NFKBIB detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. IKB BETA/NFKBIB information: Molecular Weight: 37771 MW; Subcellular Localization: Cytoplasm . Nucleus ; Tissue Specificity: Expressed in all tissues examined.
Type: Primary
Antigen: IKB Beta
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat