Order Entry
Sweden
ContactUsLinkComponent
Anti-HOXA9 Polyclonal Antibody
Anti-HOXA9 Polyclonal Antibody
  BSBTPB9216
undefined
Anti-HOXA9 Polyclonal Antibody
  BSBTPB9216

 

  • Antibody type:
    Primary
  • Antigen symbol:
    HOXA9
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Rat
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Polyclonal antibody for HOXA9 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Human. HOXA9 information: Molecular Weight: 30172 MW; Subcellular Localization: Nucleus.

Type: Primary
Antigen: HOXA9
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Rat