Order Entry
Sweden
ContactUsLinkComponent
Anti-GRIK1 Polyclonal Antibody
Anti-GRIK1 Polyclonal Antibody
Catalog # BSBTPB9208
Supplier:  Boster Bio
Anti-GRIK1 Polyclonal Antibody
Catalog # BSBTPB9208
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    GRIK1
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Polyclonal antibody for GRIK1 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Human. GRIK1 information: Molecular Weight: 103981 MW; Subcellular Localization: Cell membrane; Multi-pass membrane protein. Cell junction, synapse, postsynaptic cell membrane; Multi-pass membrane protein; Tissue Specificity: Most abundant in the cerebellum and the suprachiasmatic nuclei (SCN) of the hypothalamus.

Type: Primary
Antigen: GRIK1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat