Anti-GATA3 Polyclonal Antibody
Catalog # BSBTPB9199
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen symbol:GATA3
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human,Rat
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Polyclonal antibody for GATA-3/GATA3 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Human. GATA-3/GATA3 information: Molecular Weight: 47916 MW; Subcellular Localization: Nucleus; Tissue Specificity: T-cells and endothelial cells.
Type: Primary
Antigen: GATA3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Rat