Order Entry
Sweden
ContactUsLinkComponent
Anti-Fos B Polyclonal Antibody
Anti-Fos B Polyclonal Antibody
Catalog # BSBTRP1086
Supplier:  Boster Bio
undefined
Anti-Fos B Polyclonal Antibody
Catalog # BSBTRP1086
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    Fos B
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Polyclonal antibody for G0S3/FOSB detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. G0S3/FOSB information: Molecular Weight: 35928 MW; Subcellular Localization: Nucleus.

Type: Primary
Antigen: Fos B
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat