Anti-Eotaxin Polyclonal Antibody
Catalog # BSBTPB9065
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen symbol:Eotaxin
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Polyclonal antibody for Eotaxin/CCL11 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Mouse. Eotaxin/CCL11 information: Molecular Weight: 10893 MW; Subcellular Localization: Secreted; Tissue Specificity: Expressed constitutively in the thymus. Expression inducible in the lung (type I alveolar epithelial cells), intestine, heart, spleen, kidney.
Type: Primary
Antigen: Eotaxin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Mouse