Anti-EAAT2 Polyclonal Antibody
BSBTRP1065
- Antibody type:Primary
- Antigen symbol:EAAT2
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Polyclonal antibody for EAAT2/SLC1A2 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Human. EAAT2/SLC1A2 information: Molecular Weight: 62104 MW; Subcellular Localization: Membrane; Multi-pass membrane protein.
Type: Primary
Antigen: EAAT2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat