Anti-complement C5a Polyclonal Antibody
BSBTPA2308
- Antibody type:Primary
- Antigen symbol:complement C5a
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Polyclonal antibody for C5A/C5 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Mouse. C5A/C5 information: Molecular Weight: 8981 MW; Subcellular Localization: Secreted.
Type: Primary
Antigen: complement C5a
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Mouse; Rat