Anti-CMA1/Chymase Polyclonal Antibody
BSBTPB9055
- Antibody type:Primary
- Antigen symbol:CMA1/Chymase
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Polyclonal antibody for Chymase/CMA1 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. Chymase/CMA1 information: Molecular Weight: 27325 MW; Subcellular Localization: Secreted. Cytoplasmic granule. Mast cell granules; Tissue Specificity: Mast cells in lung, heart, skin and placenta. Expressed in both normal skin and in urticaria pigmentosa lesions.
Type: Primary
Antigen: CMA1/Chymase
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human