Order Entry
Sweden
ContactUsLinkComponent
Anti-CCL7/MCP-3 Polyclonal Antibody
Anti-CCL7/MCP-3 Polyclonal Antibody
Catalog # BSBTPB9029
Supplier:  Boster Bio
undefined
Anti-CCL7/MCP-3 Polyclonal Antibody
Catalog # BSBTPB9029
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    CCL7/MCP-3
  • Clonality:
    Polyclonal
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Polyclonal antibody for MCP3/CCL7 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: ELISA. Reactive species: Mouse. MCP3/CCL7 information: Molecular Weight: 10999 MW; Subcellular Localization: Secreted.

Type: Primary
Antigen: CCL7/MCP-3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Mouse