Anti-Calbindin Polyclonal Antibody
Catalog # BSBTPB9045
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Calbindin
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Polyclonal antibody for CALBINDIN/CALB1 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. CALBINDIN/CALB1 information: Molecular Weight: 30025 MW; .
Type: Primary
Antigen: Calbindin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat