Order Entry
Sweden
ContactUsLinkComponent
Anti-12 Lipoxygenase Polyclonal Antibody
Anti-12 Lipoxygenase Polyclonal Antibody
Catalog # BSBTPA1485-1
Supplier:  Boster Bio
undefined
Anti-12 Lipoxygenase Polyclonal Antibody
Catalog # BSBTPA1485-1
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    12 Lipoxygenase
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Polyclonal antibody for 12 LIPOXYGENASE/ALOX12 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Human. 12 LIPOXYGENASE/ALOX12 information: Molecular Weight: 25762 MW; Subcellular Localization: Ubiquitin: Cytoplasm . Nucleus ; Tissue Specificity: Expressed in vascular smooth muscle cells.

Type: Primary
Antigen: 12 Lipoxygenase
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human