Order Entry
Sweden
Orders LinkContactUsLinkComponent
 

Niclosamide 97+%

Environment
Niclosamide 97+%
Certificates
Niclosamide 97+%

2 Options Available Below

Product Details & Documents
  • CAS Number:50-65-7
  • Formula:C₁₃H₈Cl₂N₂O₄
  • MDL Number:MFCD00057597
  • Molecular Weight:327.12 g/mol
  • Melting Pt:225…230 °C
  • Storage Temperature:Omgivande
  • UN:3077
  • ADR:9,III
Specifications for Product Family

Specification Test Results

  • Appearance:
    Off-white to yellow Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    227 to 232 °C
  • Loss on drying:
    ≤2 % (105 °C, 4 h)
  • HPLC:
    ≥97.0 %
Product Family Options

Product Information

  • Förpackningstyp
    Förp.
    Availability
    Price
  • Glass bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 1 025,00 kr
    1 025,00 kr
    1 * 25 g
       1 025,00 kr

  • Glass bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 3 525,00 kr
    3 525,00 kr
    1 * 100 g
       3 525,00 kr