

Iodomethane 99% stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:74-88-4
:
Formula:CH₃I
MDL Number:MFCD00001073
:141.94 g/mol
Boiling Pt:42.5 °C (760 mmHg)
Melting Pt:–66 °C
Density:2.28 g/cm³
Storage Temperature:Ambient
UN:2644
ADR:6.1,I
:GHS06
:GHS08
Specification Test Results
- Appearance:Clear yellow to orange Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5290 to 1.5310 (20°C, 589 nm)
- GC:≥98.5 %
- Stabilizer:Copper