Order Entry
Sweden
ContactUsLinkComponent
8-Bromoisoquinoline 97%
Toxic
8-Bromoisoquinoline 97%
Certificates
undefined
Toxic
8-Bromoisoquinoline 97%

Specification Test Results

  • Appearance:
    White to light brown Powder or crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    74 to 82 °C
  • GC:
    ≥96.0 %

Specification Test Results