
Sodium hydroxide 2 mol/l (2 N) in aqueous solution for biopharmaceutical production
Leverantör: VWR Chemicals
Certifikat
Om denna produkt
CAS-nummer:1310-73-2
:
Formel:NaOH
MDL-nummer:MFCD00003548
:40 g/mol
Förvaringstemperatur:Ambient
EINECS:215-185-5
FN:1824
ADR:8,II
:GHS05
Specifikationstestresultat
- Assay:1.95 --> 2.05 mol/l
- Appearance:Passes test
- CO3 (as Na2CO3):Max. 1 %
- Heavy metals (as Pb):Max. 5 ppm
- Cl (Chloride):Max. 20 ppm
- SO4 (Sulphate):Max. 30 ppm
- Fe (Iron):Max. 3 ppm
- Residual metal catalysts:Unlikely to be present
- Residual solvents (ICH Q3C):Excl.by manufact. process
- Endotoxin:Max. 2.0 EU/ml
- Filtered through 0.2 μm:Confirmed
Specifikationstestresultat
Frequently Bought Together