Nitrate reagent powder pillows, NitraVer® 6
Catalog # HACH2107249
Leverantör: Hach
CAS-nummer:
Specifikationer
- Chemical method:Cadmium reduction
- Pk:100
- Description:Nitrate NitraVer® 6, LR
- Sample size:10 ml
- For unit:DR 6000; DR 3800; DR 2800; DR 2700; DR 1900; DR900
- Range:0,01 - 0,50 mg/l
Specifikationer
Om denna produkt
These reagents are for the determination of low range nitrate by the NitraVer® 6 cadmium reduction method.