Order Entry
Sweden
ContactUsLinkComponent
IPTG (isopropyl-β-D-thiogalactopyranoside) solution 100 mM, Sigma-Aldrich®
IPTG (isopropyl-β-D-thiogalactopyranoside) solution 100 mM, Sigma-Aldrich®
Catalog # 70527-3
CAS Number:  
IPTG (isopropyl-β-D-thiogalactopyranoside) solution 100 mM, Sigma-Aldrich®
Catalog # 70527-3
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    15 mL
  • Seq:
    2295

Specifications

Specification Test Results

  • Storage:
    -20 °C

Specification Test Results

About this item

 :
 :238.31 g/mol
Storage Temperature:Freezer
CAS Number:367-93-1
Formula:C₉H₁₈O₅S
MDL Number:MFCD00063273