Order Entry
Sweden
ContactUsLinkComponent
Sodium hydroxide, pellets NF, Multi-Compendial, J.T.Baker®
Frätande
Sodium hydroxide, pellets NF, Multi-Compendial, J.T.Baker®
Leverantör:  Avantor
Certifikat
undefined
Frätande
Sodium hydroxide, pellets NF, Multi-Compendial, J.T.Baker®
Leverantör:  Avantor
Certifikat

Specifikationstestresultat

  • NF - Identification A (Sodium):
    Passes Test
  • NF - Identification B (pH):
    ≥ 11.0
  • NF - Insoluble Substances and Organic Matter:
    Passes Test
  • NF - Potassium (K):
    ≤ 0.5 %
  • NF - Carbonate (as Na₂CO₃):
    ≤ 3.0 %
  • NF - Total Alkali:
    95.0 - 100.5 %
  • NF - Content of Sodium:
    54.0 - 59.8 %
  • EP/BP - Identification A:
    Passes Test
  • EP/BP - Identification B:
    Passes Test
  • EP/BP - Carbonate (as Na₂CO₃):
    ≤ 2.0 %
  • EP/BP - Appearance of Solution:
    Passes Test
  • EP/BP - Chloride (Cl):
    ≤ 200 ppm
  • EP/BP - Sulfate (SO₄):
    ≤ 200 ppm
  • EP/BP - Iron (Fe):
    ≤ 10 ppm
  • EP/BP - Assay (NaOH):
    97.0 - 100.5 %
  • JP - Assay:
    95.0 - 101.0 %
  • JP - Identification A:
    Passes Test
  • JP - Identification B:
    Passes Test
  • JP - Clarity and Color of Solution:
    Passes Test
  • JP - Chloride (Cl):
    ≤ 0.050 %
  • JP - Potassium (K):
    Passes Test
  • JP - Carbonate (as Na₂CO₃):
    ≤ 2.0 %
  • No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.:
    ≤2.0%
  • Elemental Impurities (USP 232, EP 5.20) - Information on elemental impurities for this product is available on the associated Product Regulatory Data Sheet and elemental impurity profile report.:
    Passes Test

Specifikationstestresultat

Relaterad information