Organic reference standards, Chloroparaffin C10-C13, Dr. Ehrenstorfer
Supplier: EHRENSTORFER
Certificates
About this item
- Food and environment industry applications
Specifications
- CAS No.:85422-92-0
- Storage temperature:Ambient
- Description:Chloroparaffin C10-C13 63% in Cyclohexane