Organic reference standard, cineole, Ph.Eur., RS
EDQMY0000176
: EDQM
:
- Pack type:Vial
- Pk:180 mg
- Standards:Reference standard
- CAS No.:470-82-6
- Description:Cineole
- Formula:C₁₀H₁₈O
- Storage temperature:Ambient
- Description:Cineole
- In Monograph 1973