Organic reference standard, Amisulpride, Ph.Eur., RS
Catalog # EDQMA0590000
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:10 mg
- Standards:Reference standard
- CAS No.:71675-85-9
- Description:Amisulpride
- Formula:C₁₇H₂₇N₃O₄S
- Storage temperature:Ambient
- Description:Amisulpride
Specifications
About this item
- Method: Monograph only
- API product type
- Pharmaceutical industry applications