Order Entry
Sweden
Orders LinkContactUsLinkComponent
 

Duloxetine, Reference Standards

Supplier:   USP
Duloxetine, Reference Standards
Supplier:  USP
Certificates

4 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

 : US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1229817.
  • 1229872.
  • 1229883.
  • 1229828.
  • Description:
  • Duloxetine Hydrochloride
  • Duloxetine
  • Duloxetine Related Compound H
  • Duloxetine Related Compound A
  • Synonym(s):
  • (S)-N-Methyl-3-(naphthalen-1-yloxy)-3-(thiophen-3-yl)propan-1-amine hydrochloride
  • (S)-4-{Methyl[3-(naphthalen-1-yloxy)-3-(thiophen-2-yl)propyl]amino}-4-oxobutanoic acid
  • (R)-N-Methyl-3-(naphthalen-1-yloxy)-3-(thiophen-2-yl)propan-1-amine hydrochloride
  • CAS No.:
  • 136434-34-9
  • 1104890-90-5
  • 199191-66-7
  • 910138-96-4
  • Formula:
  • C₁₈H₂₀ClNOS
  • C₁₈H₂₀ClNOS
  • C₂₂H₂₃NO₄S
  • UN No.:
  • 2811
  • 2811

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1229817.
    Duloxetine Hydrochloride
    Duloxetine Hydrochloride
    USP reference standard
    Vial
    100 mg
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 11 050,00 kr
    11 050,00 kr
    1 * 100 mg
       11 050,00 kr

    Specifications:
    More
    Specifications
    Cat. No.
    1229817.
    Description
    Duloxetine Hydrochloride
    Synonym(s)
    CAS No.
    136434-34-9
    Formula
    C₁₈H₂₀ClNOS
    UN No.
    2811
  • 1229872.
    Duloxetine
    Duloxetine
    USP reference standard
    Vial
    10 mg
    Log in for availability
    Log in for availability
    1 * 10 mg
    1 * 10 mg - 14 575,00 kr
    14 575,00 kr
    1 * 10 mg
       14 575,00 kr

    Specifications:
    More
    Specifications
    Cat. No.
    1229872.
    Description
    Duloxetine
    Synonym(s)
    (S)-N-Methyl-3-(naphthalen-1-yloxy)-3-(thiophen-3-yl)propan-1-amine hydrochloride
    CAS No.
    1104890-90-5
    Formula
    C₁₈H₂₀ClNOS
    UN No.
  • 1229883.
    Duloxetine Related Compound H
    Duloxetine Related Compound H
    USP reference standard
    Vial
    30 mg
    Log in for availability
    Log in for availability
    1 * 30 mg
    1 * 30 mg - 12 875,00 kr
    12 875,00 kr
    1 * 30 mg
       12 875,00 kr

    Specifications:
    More
    Specifications
    Cat. No.
    1229883.
    Description
    Duloxetine Related Compound H
    Synonym(s)
    (S)-4-{Methyl[3-(naphthalen-1-yloxy)-3-(thiophen-2-yl)propyl]amino}-4-oxobutanoic acid
    CAS No.
    199191-66-7
    Formula
    C₂₂H₂₃NO₄S
    UN No.
  • 1229828.
    Duloxetine Related Compound A
    Duloxetine Related Compound A
    USP reference standard
    Vial
    10 mg
    Log in for availability
    Log in for availability
    1 * 10 mg
    1 * 10 mg - 12 875,00 kr
    12 875,00 kr
    1 * 10 mg
       12 875,00 kr

    Specifications:
    More
    Specifications
    Cat. No.
    1229828.
    Description
    Duloxetine Related Compound A
    Synonym(s)
    (R)-N-Methyl-3-(naphthalen-1-yloxy)-3-(thiophen-2-yl)propan-1-amine hydrochloride
    CAS No.
    910138-96-4
    Formula
    UN No.
    2811