Order Entry
Sweden
ContactUsLinkComponent
Cromolyn Related Compounds, Reference Standard
Cromolyn Related Compounds, Reference Standard
Supplier:  USP
Certificates
Cromolyn Related Compounds, Reference Standard
Supplier:  USP

Specifications

  • Synonym(s):
    diethyl 5,5'-[(2-hydroxypropane-1,3-diyl)bis(oxy)]bis(4-oxo-4H-chromene-2-carboxylate)
  • Description:
    Cromolyn Related Compound B
  • CAS No.:
    15826-37-6
  • Formula:
    C₂₃H₁₄Na₂O₁₁

Specifications