Order Entry
Sweden
ContactUsLinkComponent
 
Clopidogrel Related Compounds, Reference Standard
 :  USP
 
Clopidogrel Related Compounds, Reference Standard
 :  USP

 

  • CAS No.:
    144750-42-5
  • Synonym(s):
    (+)-(S)-(o-chlorophenyl)-6,7-dihydrothieno[3,2-c]pyridine-5(4H)-acetic acid, hydrochloride
  • Formula:
    C₁₅H₁₅Cl₂NO₂S
  • Description:
    Clopidogrel Related Compound A
  • UN No.:
    3261