Order Entry
Sweden
ContactUsLinkComponent
Human recombinant TNF alpha (soluble) (from E. coli)
Human recombinant TNF alpha (soluble) (from E. coli)
Catalog # PRSI90-222
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant TNF alpha (soluble) (from E. coli)
Catalog # PRSI90-222
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Environmentally Preferable:
  • Tag sequence:
    DDDDK Tag
  • Storage conditions:
    Stable for at least 1 year after receipt when stored at −20 °C.
  • Biological activity:
    Induces cell death at a concentration range of 0.01-10ng/ml (ED50= 0.05ng/ml or 2 x 10E7 Units/mg; WEHI 164 cells).
  • Protein synonyms:
    Cachectin|Tumor Necrosis Factor Ligand Superfamily Member 2
  • Protein/peptide name:
    TNF alpha (soluble)
  • Purity:
    >95% (SDS-PAGE)
  • Sequence:
    85-223
  • Formulation:
    Lyophilized, Contains PBS.

Specifications

About this item

Tumour necrosis factor (TNF, cachexin or cachectin and formerly known as tumour necrosis factor-alpha) is a cytokine involved in systemic inflammation and is a member of a group of cytokines that stimulate the acute phase reaction. The receptor TNF-R1 is activated by both the membrane-bound and soluble trimeric forms of TNF-alpha, whereas the receptor TNF-R2 only responds to the membrane-bound form of TNF-alpha (MultimericTNF-alpha (Prod. No. AG-40B-0019). Since the MultimericTNF-alpha mimics the membrane-bound form (forms oligomers higher than trimer), it is the only TNF-alpha protein that can activate the TNF-R2. For TNF-R1 activation, either "normal" TNF-alpha or MultimericTNF-alpha can be used."

Fusion-Tag: DDDDK Tag

This recombinant proteins is for research use only.