- Pk:50 µG
- Protein/peptide type:Recombinant
- Species:Mouse
- Tag sequence:C-Fc tag
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Protein synonyms:TGFR-2|TGF-beta type II receptor|TGF-beta receptor type-2|Tgfbr2|TbetaR-II|Transforming growth factor-beta receptor type II|TGF-beta receptor type II
- Protein/peptide name:TGF beta Receptor II
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:42.3 kD
- Sequence:Ile24-Asp159
- Formulation:Lyophilized from a 0.2 um filtered solution of PBS,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
Transforming growth factor- beta (TGF- beta ) is an essential regulator in the processes of development, cell proliferation, and extracellular matrix deposition. TGF- beta regulates cellular processes by binding to three high-affinity cell surface receptors: TGF- beta receptor type I (TGF- beta -RI), TGF- beta receptor type II (TGF- beta -RII), and TGF- beta beta receptor type III (TGF- beta -RIII). TGF- beta RII is consists of a C-terminal protein kinase domain and an N-terminal ectodomain and belongs to transforming growth factor-beta (TGF- beta ) receptor subfamily. TGF- beta RII has a protein kinase domain which can form a heterodimeric complex with another receptor protein and bind TGF-beta. This receptor/ligand complex phosphorylates protein will enter the nucleus and regulate the transcription of a subset of genes related to cell proliferation.
Fusion-Tag: C-Fc tag
This recombinant protein can be used for biological assays. For research use only.