Order Entry
Sweden
ContactUsLinkComponent
Human recombinant Ferritin heavy chain (from E. coli)
Human recombinant Ferritin heavy chain (from E. coli)
Catalog # PRSI92-267
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant Ferritin heavy chain (from E. coli)
Catalog # PRSI92-267
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    N-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    Ferritin heavy chain|FTH1|FTH|Cell proliferation-inducing gene 15 protein|Ferritin H subunit|FTHL6
  • Protein/peptide name:
    Ferritin heavy chain
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    23.4 kD
  • Sequence:
    Met1-Ser183
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Ferritin heavy polypeptide 1(FTH1), is a ubiquitous intracellular protein which stores iron in a soluble, non-toxic, readily available form. FTH1 has ferroxidase activity and is important for iron homeostasis. Iron is taken up in the ferrous form and deposited as ferric hydroxides after oxidation. Ferritin is composed of 24 subunits of the light and heavy ferritin chains. It plays a role in delivery of iron to cells and mediates iron uptake in capsule cells of the developing kidney. Variation of ferritin subunit composition may affect iron absorption and release in different tissues. Deficiency of ferritin proteins may cause several neurodegenerative diseases. Almost all living organisms can produce this protein, including algae, bacteria, higher plants, and animals.

Fusion-Tag: N-6 His tag

This recombinant protein can be used for biological assays. For research use only.