Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:Cells
- Species:Human
- Tag sequence:C-Fc tag
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Protein synonyms:LAMP1|CD107a|Lysosome-Associated Membrane Protein 1|Lysosome-Associated Membrane Glycoprotein 1|LAMP-1|CD107 Antigen-like Family Member A
- Protein/peptide name:LAMP1
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:65.5 kD
- Sequence:Ala29-Met382
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM PB,150mM NaCl,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:used for biological assays
Specifications
About this item
Lysosome-Associated Membrane Glycoprotein 1 (LAMP1) is a single-pass type I membrane protein belonging to the LAMP family. LAMP1 is expressed largely in the endosome-lysosome membranes of cells.It shuttles between lysosomes, endosomes, and the plasma membrane. LAMP1 functions to present carbohydrate ligands to selectins and it has also been implicated in tumor cell metastasis. It has been proposed LAMP1 can be used as a therapeutic agent for certain cancers, as well as a marker for lysosomal storage disorders and degranulation on lymphocytes such as CD8+ and NK cells. Cell surface LAMP1 and LAMP2 have been shown to promote adhesion of human peripheral blood mononuclear cells(PBMC) to vascular endothelium, therefore they are possibly involved in the adhesion of PBMCs to the site of inflammation.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-Fc tag