Human recombinant Annexin A1 (from E. coli)
Katalog # PRSI91-076
Leverantör: ProSci Inc.
CAS-nummer:
Specifikationer
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Protein synonyms:Lipocortin I|Annexin I|Chromobindin-9|Calpactin II|p35|Calpactin-2|ANXA1|Annexin-1|Phospholipase A2 Inhibitory Protein|Annexin A1
- Protein/peptide name:Annexin A1
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:38.58 kD
- Sequence:Ala2-Asn346
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifikationer
Om denna produkt
Annexin A1 is the first characterized member of the annexin family of proteins and is able to bind to cellular membranes in a calcium-dependent manner, promoting membrane fusion and endocytosis. Annexin A1 has anti-inflammatory properties and inhibits phospholipase A2 activity. Annexin A1 also has roles in many diverse cellular functions, such as membrane aggregation, inflammation, phagocytosis, proliferation, apoptosis, and tumorigenesis and cancer development. ANXA1 is strongly expressed on the cell membrane and occasionally in the cytoplasm of tumor cells in 97% of samples from patients with hairy cell leukemia.
- This recombinant protein can be used for biological assays. For research use only.